Acetamidine Hydroiodide, low water content ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Acetamidinium iodide
A2902-1G
A2902-5G
TCIAA2902-1GEA
55
EUR
TCIAA2902-1G
TCIAA2902-5G
Acetamidine Hydroiodide, low water content ≥98.0% (by titrimetric analysis)
Acetamidine Hydroiodide
Formula:
C₂H₆N₂·HI Peso molecolare: 186,00 g/mol |
CAS Number:
1452099-14-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Water | max. 100 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...