Stampa…

2-Amino-5-iodo-4-methylpyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Iodo-4-methyl-pyridin-2-ylamine, 5-Iodo-4-methylpyridin-2-ylamine

A2880-1G A2880-5G
TCIAA2880-1GEA 33 EUR
TCIAA2880-1G TCIAA2880-5G
2-Amino-5-iodo-4-methylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-iodo-4-methylpyridine
Formula: C₆H₇IN₂
Peso molecolare: 234,04 g/mol
MDL Number: MFCD06637727
CAS Number: 356561-08-5

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 119.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR