2-Amino-6-fluoropyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
A2879-1G
A2879-5G
TCIAA2879-1GEA
32
EUR
TCIAA2879-1G
TCIAA2879-5G
2-Amino-6-fluoropyridine ≥98.0% (by GC, titration analysis)
2-Amino-6-fluoropyridine
Formula:
C₅H₅FN₂ Peso molecolare: 112,11 g/mol |
MDL Number:
MFCD04114200 CAS Number: 1597-32-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...