Stampa…

Ancymidol ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: alpha-Cyclopropyl-alpha-(4-methoxyphenyl)-5-pyrimidinemethanol

A2877-25MG A2877-100MG
TCIAA2877-25MGEA 43 EUR
TCIAA2877-25MG TCIAA2877-100MG
Ancymidol ≥98.0% (by GC)
Ancymidol
Formula: C₁₅H₁₆N₂O₂
Peso molecolare: 256,31 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00072501
CAS Number: 12771-68-5
EINECS: 235-814-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 109.0 to 113.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR