3-Amino-5-chloropyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Chloropyridin-3-amine
A2876-1G
A2876-5G
TCIAA2876-1GEA
60
EUR
TCIAA2876-1G
TCIAA2876-5G
3-Amino-5-chloropyridine ≥98.0% (by GC, titration analysis)
3-Amino-5-chloropyridine
Formula:
C₅H₅ClN₂ Peso molecolare: 128,56 g/mol |
MDL Number:
MFCD03701386 CAS Number: 22353-34-0 |
Risultati del test di specificazione
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...