Stampa…

Abiraterone ≥98.0% (by HPLC)

Fornitore: TCI
Attenzione

Synonyms: CB 7598, CB-7598, CYP17 inhibitor, CB7598 (3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-pyridin-3-yl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-ol

A2868-20MG A2868-200MG
TCIAA2868-20MGEA 19 EUR
TCIAA2868-20MG TCIAA2868-200MG
Abiraterone ≥98.0% (by HPLC)
Abiraterone
Formula: C₂₄H₃₁NO
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00924100
CAS Number: 154229-19-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 226.0 to 230.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR