Stampa…

5-Acetylthiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A2866-1G A2866-5G
TCIAA2866-1GEA 21 EUR
TCIAA2866-1G TCIAA2866-5G
5-Acetylthiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Acetylthiophene-2-carboxylic acid
Formula: C₇H₆O₃S
Peso molecolare: 170,19 g/mol
MDL Number: MFCD00055512
CAS Number: 4066-41-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 209.0 to 213.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR