1-Acryloyloxy-3-hydroxyadamantane ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Acrylic acid 3-Hydroxy-1-adamantyl ester, 3-Hydroxyadamantan-1-yl acrylate, 3-Hydroxy-1-adamantyl acrylate
A2859-5G
A2859-25G
TCIAA2859-5GEA
63
EUR
TCIAA2859-5G
TCIAA2859-25G
1-Acryloyloxy-3-hydroxyadamantane ≥98.0% (by GC)
1-Acryloyloxy-3-hydroxyadamantane
Formula:
C₁₃H₁₈O₃ Peso molecolare: 222,28 g/mol |
MDL Number:
MFCD13194929 CAS Number: 216581-76-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...