2-Amino-4,5-dimethylbenzoic acid ≥97.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,5-Dimethylanthranilic acid
A2850-200MG
A2850-1G
TCIAA2850-200MGEA
44
EUR
TCIAA2850-200MG
TCIAA2850-1G
2-Amino-4,5-dimethylbenzoic acid ≥97.0% (by HPLC, titration analysis)
2-Amino-4,5-dimethylbenzoic acid
Formula:
C₉H₁₁NO₂ Peso molecolare: 165,19 g/mol |
MDL Number:
MFCD00156998 CAS Number: 15089-51-7 EINECS: 247-201-1 |
Risultati del test di specificazione
Appearance | White to Yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...