Stampa…

2-Amino-4-phenylthiazole ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Phenylthiazol-2-amine, 4-Phenyl-1,3-thiazol-2-amine

A2846-1G A2846-5G
TCIAA2846-1GEA 47 EUR
TCIAA2846-1G TCIAA2846-5G
2-Amino-4-phenylthiazole ≥98.0% (by GC, titration analysis)
2-Amino-4-phenylthiazole
Formula: C₉H₈N₂S
Peso molecolare: 176,24 g/mol
MDL Number: MFCD00039680
CAS Number: 2010-06-2
EINECS: 217-926-8
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR