Stampa…

trans-4-Acetoxycinnamic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (E)-3-(4-Acetoxyphenyl)acrylic acid, trans-3-(4-Acetoxyphenyl)-2-propenoic acid

A2845-5G A2845-25G
TCIAA2845-5GEA 103 EUR
TCIAA2845-5G TCIAA2845-25G
trans-4-Acetoxycinnamic acid ≥98.0% (by GC, titration analysis)
trans-4-Acetoxycinnamic acid
Formula: C₁₁H₁₀O₄
Peso molecolare: 206,20 g/mol
MDL Number: MFCD00016847
CAS Number: 27542-85-4
EINECS: 239-512-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 208.0 to 212.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR