5-Amino-2,3-dichloropyridine ≥98.0% (by GC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,3-Dichloropyridin-5-amine, 2,3-Dichloro-5-pyridylamine, 3-Amino-5,6-dichloropyridine
A2824-1G
A2824-5G
TCIAA2824-1GEA
54
EUR
TCIAA2824-1G
TCIAA2824-5G
5-Amino-2,3-dichloropyridine ≥98.0% (by GC, total nitrogen)
5-Amino-2,3-dichloropyridine
Formula:
C₅H₄Cl₂N₂ Peso molecolare: 163,01 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD03840434 CAS Number: 98121-41-6 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 111.0 to 115.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...