Stampa…

Isoquinoline-1-amine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Isoquinolin-1-amine, 1-Aminoisoquinoline

A2817-1G A2817-5G
TCIAA2817-1GEA 50 EUR
TCIAA2817-1G TCIAA2817-5G
Isoquinoline-1-amine ≥98.0% (by GC)
Isoquinoline-1-amine
Formula: C₉H₈N₂
Peso molecolare: 144,18 g/mol
Punto di ebollizione: 166°C
MDL Number: MFCD00024137
CAS Number: 1532-84-9
EINECS: 216-243-2

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 119.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR