Stampa…
3-Cyano-4-methylaniline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Pericolo
Synonyms: 5-Amino-2-methylbenzonitrile
A2814-1G A2814-5G
TCIAA2814-1GEA 16 EUR
TCIAA2814-1G TCIAA2814-5G
3-Cyano-4-methylaniline ≥98.0% (by GC, titration analysis)
3-Cyano-4-methylaniline
Formula: C₈H₈N₂
Peso molecolare: 132,16 g/mol
Punto di ebollizione: 110°C
MDL Number: MFCD00017608
CAS Number: 50670-64-9
UN: 3439
ADR: 6.1,III
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 89.0 to 93.0 deg-C