Stampa…

Aminophylline ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 3,7-Dihydro-1,3-dimethylpurine-2,6-dione, complex with 1,2-ethanediamine (2:1), 1,3-Dimethyl-3,7-dihydro-1H-purine-2,6-dione hemiethane-1,2-diamine salt

A2805-25G A2805-100G
TCIAA2805-25GEA 27 EUR
TCIAA2805-25G TCIAA2805-100G
Aminophylline ≥98.0% (by HPLC)
Aminophylline
Formula: C₇H₈N₄O₂
Peso molecolare: 420,43 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00013221
CAS Number: 317-34-0
EINECS: 206-264-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,00465

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR