3-Amino-3-methyloxetane ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Methyloxetan-3-amine, 3-Methyl-3-oxetanamine
A2792-200MG
A2792-1G
TCIAA2792-200MGEA
81
EUR
TCIAA2792-200MG
TCIAA2792-1G
3-Amino-3-methyloxetane ≥97.0% (by GC, titration analysis)
3-Amino-3-methyloxetane
Formula:
C₄H₉NO Peso molecolare: 87,12 g/mol Flash Pt: 97°C |
MDL Number:
MFCD11111713 CAS Number: 874473-14-0 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...