2-Amino-5-chloropyrazine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
A2790-1G
A2790-5G
TCIAA2790-1GEA
37
EUR
TCIAA2790-1G
TCIAA2790-5G
2-Amino-5-chloropyrazine ≥98.0% (by GC, titration analysis)
2-Amino-5-chloropyrazine
Formula:
C₄H₄ClN₃ Peso molecolare: 129,55 g/mol |
MDL Number:
MFCD03423596 CAS Number: 33332-29-5 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...