Stampa…

3-Aminofluoranthene ≥96.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Fluoranthen-3-amine

A2789-1G A2789-5G
TCIAA2789-1GEA 68 EUR
TCIAA2789-1G TCIAA2789-5G
3-Aminofluoranthene ≥96.0% (by GC, titration analysis)
3-Aminofluoranthene
Formula: C₁₆H₁₁N
Peso molecolare: 217,2700 g/mol
MDL Number: MFCD00001185
CAS Number: 2693-46-1
EINECS: 220-263-7
UN: 3077
ADR: 9,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Amber powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR