(R)-(-)-3-Aminopiperidine dihydrochloride ≥97.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₅H₁₄Cl₂N₂ |
MDL Number:
MFCD06799458 CAS Number: 334618-23-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 95.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 206.0 to 210.0 deg-C |
Specific rotation [a]20/D | -1.0 to -3.0 deg (C=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...