Stampa…

(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) ) ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), 7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, (7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium Hexafluorophosphate, AOP, Tris(dimethylamino)(3H-1,2,3-triazolo[4,5-b]pyridin-3-yloxy)phosphonium Hexafluorophosphate

A2782-1G A2782-5G
TCIAA2782-1GEA 38 EUR
TCIAA2782-1G TCIAA2782-5G
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) ) ≥98.0% (by HPLC)
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) )
Formula: C₁₂H₂₂F₆N₆OP₂
Peso molecolare: 443,27 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD09263289
CAS Number: 156311-85-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Lambda max. 245.0 to 248.0 nm (CH3CN)
Absorbance min. 0.280 (0.02g/L, CH3CN, 245.0 to 248.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR