Stampa…

2-Acetamido-6-methylpyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-(6-Methylpyridin-2-yl)acetamide

A2780-5G A2780-25G
TCIAA2780-5GEA 56 EUR
TCIAA2780-5G TCIAA2780-25G
2-Acetamido-6-methylpyridine ≥98.0% (by GC)
2-Acetamido-6-methylpyridine
Formula: C₈H₁₀N₂O
Peso molecolare: 150,18 g/mol
MDL Number: MFCD00091881
CAS Number: 5327-33-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR