Stampa…

Amsacrine hydrochloride ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: m-AMSA, N-(4-(Acridin-9-ylamino)-3-methoxyphenyl)methanesulphonamide hydrochloride, 4-(9-Acridinylamino)-N-(methanesulphonyl)-m-anisidine hydrochloride, Amsacrine HCl

A2777-20MG A2777-100MG
TCIAA2777-20MGEA 42 EUR
TCIAA2777-20MG TCIAA2777-100MG
Amsacrine hydrochloride ≥98.0% (by HPLC)
Amsacrine hydrochloride
Formula: C₂₁H₁₉N₃O₃S·HCl
Peso molecolare: 429,92 g/mol
Punto di fusione: 288°C
MDL Number: MFCD07799963
CAS Number: 54301-15-4
UN: 2811
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 9.20 to 10.20 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR