Stampa…

β-Alanine-tert-butylester hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI

A2772-5G A2772-25G
TCIAA2772-5GEA 49 EUR
TCIAA2772-5G TCIAA2772-25G
β-Alanine-tert-butylester hydrochloride ≥98.0% (by titrimetric analysis)
beta-Alanine-tert-butylester hydrochloride
Formula: C₇H₁₆ClNO₂
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00058258
CAS Number: 58620-93-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR