Stampa…

L-Alanine benzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: H-Ala-OBzl.HCl, H-Ala-OBzl·HCl

A2771-5G A2771-25G
TCIAA2771-5GEA 20 EUR
TCIAA2771-5G TCIAA2771-25G
L-Alanine benzyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
L-Alanine benzyl ester hydrochloride
Formula: C₁₀H₁₃NO₂
Peso molecolare: 215,68 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00054340
CAS Number: 5557-83-5
EINECS: 226-920-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C
Specific rotation [a]20/D -9.0 to -11.0 deg (C=1,1mol/L HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR