Stampa…

(Acetylacetonato)(norbornadiene)rhodium(I)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (Acetylacetonato)[bicyclo[2.2.1]hepta-2,5-diene]rhodium(I), (Norbornadiene)rhodium(I) 2,4-pentanedionate

A2761-200MG A2761-1G
TCIAA2761-200MGEA 149 EUR
TCIAA2761-200MG TCIAA2761-1G
(Acetylacetonato)(norbornadiene)rhodium(I)
(Acetylacetonato)(norbornadiene)rhodium(I)
Formula: C₁₂H₁₅O₂Rh
Peso molecolare: 294,16 g/mol
MDL Number: MFCD00075051
CAS Number: 32354-50-0

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Elemental analysis (Carbon) 47.00 to 51.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR