Stampa…

5-Amino-2-(4-aminophenyl)benzimidazole ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(4-Aminophenyl)-1H-benzimidazol-5-amine

A2759-1G A2759-5G
TCIAA2759-1GEA 20 EUR
TCIAA2759-1G TCIAA2759-5G
5-Amino-2-(4-aminophenyl)benzimidazole ≥98.0% (by HPLC)
5-Amino-2-(4-aminophenyl)benzimidazole
Formula: C₁₃H₁₂N₄
Peso molecolare: 224,27 g/mol
MDL Number: MFCD00451475
CAS Number: 7621-86-5
EINECS: 231-538-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR