1-N-Cbz-4-aminopiperidine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Amino-1-piperidinecarboxylic acid Benzyl ester, 4-Amino-1-benzyloxycarbonylpiperidine, 4-Amino-1-carbobenzoxypiperidine, Benzyl 4-aminopiperidine-1-carboxylate, 4-Amino-1-Cbz-piperidine, 1-N-Cbz-4-Aminopiperidine, Benzyl 4-Amino-1-piperidinecarboxylate
Formula:
C₁₃H₁₈N₂O₂ Peso molecolare: 234,30 g/mol |
MDL Number:
MFCD05863884 CAS Number: 120278-07-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...