Stampa…

Arabinofuranosyl-guanine hydrate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Guanine 9-β-D-Arabinofuranoside, Araguanosine, Ara-G

A2739-10MG A2739-50MG
TCIAA2739-10MGEA 16 EUR
TCIAA2739-10MG TCIAA2739-50MG
Arabinofuranosyl-guanine hydrate ≥98.0% (by HPLC)
Arabinofuranosyl-guanine hydrate
Formula: C₁₀H₁₃N₅O₅
Peso molecolare: 283,24 g/mol
Punto di fusione: 225°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00065486
CAS Number: 38819-10-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 24.00% to 25.50% (calcd.on anh.substance)
Water 4.0 to 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR