Stampa…

4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: PP 3, 1-Phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine

A2737-200MG A2737-1G
TCIAA2737-200MGEA 34 EUR
TCIAA2737-200MG TCIAA2737-1G
4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine ≥98.0% (by HPLC)
4-Amino-1-phenylpyrazolo[3,4-d]pyrimidine
Formula: C₁₁H₉N₅
Peso molecolare: 211,23 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00022901
CAS Number: 5334-30-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR