Stampa…

3-Acetyl-6-bromo-2H-chromen-2-one ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Acetyl-6-bromocoumarin, 3-Acetyl-6-bromo-2H-chromen-2-one

A2723-1G A2723-5G
TCIAA2723-1GEA 39 EUR
TCIAA2723-1G TCIAA2723-5G
3-Acetyl-6-bromo-2H-chromen-2-one ≥96.0% (by GC)
3-Acetyl-6-bromo-2H-chromen-2-one
Formula: C₁₁H₇BrO₃
Peso molecolare: 267,09 g/mol
MDL Number: MFCD00024075
CAS Number: 2199-93-1

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 223.0 to 227.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR