Stampa…

5-Amino-4,6-dichloro-2-(propylthio)pyrimidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4,6-Dichloro-2-(propylthio)pyrimidin-5-amine

A2716-1G A2716-5G
TCIAA2716-1GEA 44 EUR
TCIAA2716-1G TCIAA2716-5G
5-Amino-4,6-dichloro-2-(propylthio)pyrimidine ≥98.0% (by GC)
5-Amino-4,6-dichloro-2-(propylthio)pyrimidine
Formula: C₇H₉Cl₂N₃S
Peso molecolare: 238,13 g/mol
MDL Number: MFCD16875503
CAS Number: 145783-15-9

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR