5'-Acetamido-2'-hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₀H₁₁NO₃ |
MDL Number:
MFCD00100490 CAS Number: 7298-67-1 EINECS: 230-735-4 |
Risultati del test di specificazione
Appearance | White to Light yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 166.0 to 170.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...