Stampa…

Abacavir ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (1S,4R)-4-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]-2-cyclopentene-1-methanol, ((1S,4R)-4-(2-Amino-6-(cyclopropylamino)-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol

A2694-1G A2694-5G
TCIAA2694-1GEA 79 EUR
TCIAA2694-1G TCIAA2694-5G
Abacavir ≥98.0% (by HPLC, titration analysis)
Abacavir
Formula: C₁₄H₁₈N₆O
Peso molecolare: 286,34 g/mol
MDL Number: MFCD00903850
CAS Number: 136470-78-5
Merck Index: 14,00001

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -53.0 to -56.0 deg (C=0.15, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR