Stampa…

Aluminium bromide, anidro ≥95.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

A2692-5G A2692-25G
TCIAA2692-5GEA 30 EUR
TCIAA2692-5G TCIAA2692-25G
Aluminium bromide, anidro ≥95.0% (by titrimetric analysis)
Aluminium bromide
Formula: AlBr₃
Punto di ebollizione: 256°C
Punto di fusione: 98°C
MDL Number: MFCD00003421
CAS Number: 7727-15-3
EINECS: 231-779-7
UN: 1725
ADR: 8,II
Merck Index: 14,00332

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (Chelometric Titration) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR