Stampa…

2-Amino-5-iodo-3-methylpyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Iodo-3-methylpyridin-2-amine

A2683-1G A2683-5G
TCIAA2683-1GEA 81 EUR
TCIAA2683-1G TCIAA2683-5G
2-Amino-5-iodo-3-methylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-iodo-3-methylpyridine
Formula: C₆H₇IN₂
Peso molecolare: 234,04 g/mol
MDL Number: MFCD02102422
CAS Number: 166266-19-9

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR