2-Amino-5-bromo-3-pyridinol ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Amino-5-bromo-3-hydroxypyridine
A2673-1G
A2673-5G
TCIAA2673-1GEA
23
EUR
TCIAA2673-1G
TCIAA2673-5G
2-Amino-5-bromo-3-pyridinol ≥98.0% (by HPLC)
2-Amino-5-bromo-3-pyridinol
Formula:
C₅H₅BrN₂O Peso molecolare: 189,01 g/mol |
MDL Number:
MFCD09744143 CAS Number: 39903-01-0 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...