5-Aminooxindole ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Aminoindolin-2-one, 5-Amino-1,3-dihydro-indol-2-one
A2668-1G
A2668-5G
TCIAA2668-1GEA
85
EUR
TCIAA2668-1G
TCIAA2668-5G
5-Aminooxindole ≥98.0% (by HPLC, titration analysis)
5-Aminooxindole
Formula:
C₈H₈N₂O Peso molecolare: 148,16 g/mol |
MDL Number:
MFCD02179603 CAS Number: 20876-36-2 |
Risultati del test di specificazione
Appearance | White to Brown powder to crystalline |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 196.0 to 200.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...