Stampa…

2-Amino-4-(p-tolyl)thiazole ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(p-Tolyl)-2-thiazolamine, 4-(4-Methylphenyl)-2-thiazolamine, 4-(p-Tolyl)thiazol-2-amine, 2-Amino-4-(4-methylphenyl)thiazole

A2666-5G A2666-25G
TCIAA2666-5GEA 58 EUR
TCIAA2666-5G TCIAA2666-25G
2-Amino-4-(p-tolyl)thiazole ≥98.0% (by GC, titration analysis)
2-Amino-4-(p-tolyl)thiazole
Formula: C₁₀H₁₀N₂S
Peso molecolare: 190,26 g/mol
MDL Number: MFCD00170264
CAS Number: 2103-91-5

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 135.0 to 139.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR