Anthracene-9,10-dicarboxaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
9,10-Diformylanthracene, Anthracene-9,10-dicarbaldehyde
A2664-1G
A2664-5G
TCIAA2664-1GEA
110
EUR
TCIAA2664-1G
TCIAA2664-5G
Anthracene-9,10-dicarboxaldehyde ≥98.0% (by GC)
Anthracene-9,10-dicarboxaldehyde
Formula:
C₁₆H₁₀O₂ Peso molecolare: 234,25 g/mol |
MDL Number:
MFCD00045386 CAS Number: 7044-91-9 EINECS: 230-322-9 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 242.0 to 246.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...