Stampa…

DL-2-Allylglycine ≥98.0% (by HPLC)

Fornitore: TCI
Attenzione

Synonyms: DL-2-Amino-4-pentenoic acid, DL-2-Allylglycine, 2-Amino-4-pentenoic Acid, 2-Aminopent-4-enoic acid

A2663-1G A2663-5G
TCIAA2663-1GEA 65 EUR
TCIAA2663-1G TCIAA2663-5G
DL-2-Allylglycine ≥98.0% (by HPLC)
DL-2-Allylglycine
Formula: C₅H₉NO₂
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00063103
CAS Number: 7685-44-1
EINECS: 231-689-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR