4-Amino-3-iodopyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Iodopyridin-4-amine, 3-Iodo-4-pyridylamine
A2641-1G
A2641-5G
TCIAA2641-1GEA
38
EUR
TCIAA2641-1G
TCIAA2641-5G
4-Amino-3-iodopyridine ≥98.0% (by GC, titration analysis)
4-Amino-3-iodopyridine
Formula:
C₅H₅IN₂ Peso molecolare: 220,01 g/mol |
MDL Number:
MFCD02663853 CAS Number: 88511-27-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 96.0 to 100.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...