3-Aminopropyldimethoxymethylsilane ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
3-(Dimethoxymethylsilyl)propylamine
A2628-25ML
A2628-100ML
TCIAA2628-25MLEA
32
EUR
TCIAA2628-25ML
TCIAA2628-100ML
3-Aminopropyldimethoxymethylsilane ≥97.0% (by GC, titration analysis)
3-Aminopropyldimethoxymethylsilane
Formula:
C₆H₁₇NO₂Si Peso molecolare: 163,29 g/mol Punto di ebollizione: 84°C |
MDL Number:
MFCD00025614 CAS Number: 3663-44-3 EINECS: 222-919-8 UN: 3267 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...