Stampa…

4-(4-Aminophenoxy)-N-methylpicolinamide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(4-Aminophenoxy)-N-methylpicolinamide, 4-(4-Aminophenoxy)-N-methyl-2-pyridinecarboxamide

A2617-1G A2617-5G
TCIAA2617-1GEA 29 EUR
TCIAA2617-1G TCIAA2617-5G
4-(4-Aminophenoxy)-N-methylpicolinamide ≥98.0% (by HPLC, titration analysis)
4-(4-Aminophenoxy)-N-methylpicolinamide
Formula: C₁₃H₁₃N₃O₂
Peso molecolare: 243,2612 g/mol
MDL Number: MFCD08692047
CAS Number: 284462-37-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR