4-(2-Aminoethyl)cyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥97.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₈H₁₈N₂ Punto di ebollizione: 129°C |
CAS Number:
202256-86-8 UN: 2735 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % (total of isomers) |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...