Stampa…

2-Amino-N-methylbenzamide ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

A2597-1G A2597-5G
TCIAA2597-1GEA 57 EUR
TCIAA2597-1G TCIAA2597-5G
2-Amino-N-methylbenzamide ≥98.0% (by GC, titration analysis)
2-Amino-N-methylbenzamide
Formula: C₈H₁₀N₂O
MDL Number: MFCD00060602
CAS Number: 4141-08-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 77.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR