Stampa…

2-Amino-3-bromo-5-methylbenzoic acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

A2569-1G A2569-5G
TCIAA2569-1GEA 20 EUR
TCIAA2569-1G TCIAA2569-5G
2-Amino-3-bromo-5-methylbenzoic acid ≥97.0% (by titrimetric analysis)
2-Amino-3-bromo-5-methylbenzoic acid
Formula: C₈H₈BrNO₂
Peso molecolare: 230,06 g/mol
MDL Number: MFCD00051705
CAS Number: 13091-43-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystalline
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR