Stampa…

1-Amino-2-methylpyridinium iodide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A2558-5G A2558-1G
TCIAA2558-5GEA 210 EUR
TCIAA2558-5G TCIAA2558-1G
1-Amino-2-methylpyridinium iodide ≥98.0% (by HPLC, titration analysis)
1-Amino-2-methylpyridinium iodide
Formula: C₆H₉IN₂
Peso molecolare: 236,06 g/mol
MDL Number: MFCD00955862
CAS Number: 7583-90-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR