Stampa…

4-(Aminomethyl)tetrahydropyran ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (Tetrahydro-2H-pyran-4-yl)methanamine

A2550-1G A2550-5G
TCIAA2550-1GEA 40 EUR
TCIAA2550-1G TCIAA2550-5G
4-(Aminomethyl)tetrahydropyran ≥98.0% (by GC, titration analysis)
4-(Aminomethyl)tetrahydropyran
Formula: C₆H₁₃NO
Peso molecolare: 115,1735 g/mol
Punto di ebollizione: 76°C
MDL Number: MFCD02179435
CAS Number: 130290-79-8
UN: 2735
ADR: 8,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR