Stampa…

Atovaquone ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

A2545-200MG A2545-1G
TCIAA2545-200MGEA 42 EUR
TCIAA2545-200MG TCIAA2545-1G
Atovaquone ≥98.0% (by HPLC, titration analysis)
Atovaquone
Formula: C₂₂H₁₉ClO₃
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00889188
CAS Number: 95233-18-4
Merck Index: 14,00866

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 221.0 to 225.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR