2-Amino-3-iodopyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
A2542-1G
A2542-5G
TCIAA2542-1GEA
47
EUR
TCIAA2542-1G
TCIAA2542-5G
2-Amino-3-iodopyridine ≥98.0% (by GC, titration analysis)
2-Amino-3-iodopyridine
Formula:
C₅H₅IN₂ Peso molecolare: 220,01 g/mol |
MDL Number:
MFCD01651885 CAS Number: 104830-06-0 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...